Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Saraf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026432.3 | NP_080708.3 | 4 | 960 | ||
GenBank mRNA | AK007787.1 | - | 4 | 887 | ||
AK075744.1 | - | 4 | 851 | |||
AK077118.1 | - | 4 | 947 | |||
AK088728.1 | - | 4 | 843 | |||
AK093942.1 | - | 4 | 935 | |||
AK163218.1 | - | 4 | 893 | |||
AK206196.1 | - | 2 | 131 | |||
BC013497.1 | - | 4 | 901 | 4222385 | ||
BC022616.1 | - | 4 | 938 | 4187981 | ||
BC024888.1 | - | 4 | 896 | 3988822 |