Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026443.4 | NP_080719.2 | 3, 4 | 578 | ||
GenBank mRNA | AK006148.1 | - | 3, 4 | 499 | ||
AK010971.1 | - | 3, 4 | 535 | |||
AK012058.1 | - | 3, 4 | 545 | |||
AK078381.1 | - | 3, 4 | 579 | |||
BC019205.1 | - | 3, 4 | 588 | |||
BC058737.1 | - | 3, 4 | 528 | 5400435 |