Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018771.3 | NP_061241.1 | 2, 3 | 381 | ||
GenBank mRNA | AF061263.1 | - | 1, 2 | 315 | ||
AF104358.1 | - | 2, 3 | 359 | |||
AK002492.1 | - | 2, 3 | 362 | |||
AK086506.1 | - | 2, 3 | 382 | |||
AK183407.1 | - | 1, 2 | 74 | |||
BC003490.1 | - | 2, 3 | 381 | 2811749 |