Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galntl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026449.3 | NP_080725.2 | 8 | 1256 | ||
GenBank mRNA | AK005605.1 | - | 8 | 1277 | ||
AK016415.1 | - | 8 | 1257 | |||
BC049554.1 | - | 8 | 1238 | |||
BC100447.1 | - | 8 | 1109 | |||
BC145757.1 | - | 8 | 1116 | 40131060 |