Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026452.2 | NP_080728.1 | 6, 7 | 697 | ||
GenBank mRNA | AK004527.1 | - | 2, 3 | 122 | ||
AK009179.1 | - | 6, 7 | 710 | |||
AK154343.1 | - | 6, 7 | 722 | |||
AK158979.1 | - | 6, 7 | 716 | |||
BC036386.1 | - | 6, 7 | 697 | 1067021 |