Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026454.3 | NP_080730.1 | 7, 8 | 541 | ||
GenBank mRNA | AK007549.1 | - | 7, 8 | 512 | ||
AK007937.1 | - | 7, 8 | 541 | |||
AK007960.1 | - | 7, 8 | 531 | |||
AK010957.1 | - | 8, 9 | 618 | |||
AK030594.1 | - | 521 | ||||
AK153481.1 | - | 7, 8 | 542 | |||
AK208168.1 | - | 6, 7 | 325 | |||
BC006057.1 | - | 1, 2 | 54 | |||
BC016117.1 | - | 8, 9 | 823 | 4486610 |