Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026458.4 | NP_080734.3 | 23 | 3803 | ||
GenBank mRNA | AY243470.1 | - | 23 | 3803 |