Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026474.5 | NP_080750.1 | 3 | 266 | ||
GenBank mRNA | AK007881.1 | - | 3 | 201 | ||
AK010711.1 | - | 2 | 235 | |||
AK010830.1 | - | 3 | 183 | |||
AK020433.1 | - | 3 | 185 | |||
AK045153.1 | - | 3 | 205 | |||
AK082328.1 | - | 3 | 268 | |||
AK141416.1 | - | 202 | ||||
AK148838.1 | - | 3 | 180 | |||
BC009167.1 | - | 3 | 183 | 2811924 |