Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030241.2 | NP_084517.2 | 7 | 880 | ||
GenBank mRNA | AK010593.1 | - | 6 | 879 | ||
AK010835.1 | - | 6 | 905 | |||
AK030904.1 | - | 7 | 874 | |||
AK088097.1 | - | 6 | 727 | |||
BC108333.1 | - | 7 | 809 | 4978041 |