Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Puf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164600.1 | NP_001158072.1 | 5 | 502 | ||
NM_028364.2 | NP_082640.2 | 6 | 583 | |||
NM_133691.5 | NP_598452.2 | 5 | 532 | |||
GenBank mRNA | AK012434.1 | - | 5 | 472 | ||
AK075960.1 | - | 5 | 500 | |||
AK149632.1 | - | 6 | 549 | |||
AK151526.1 | - | 5 | 496 | |||
AK168789.1 | - | 5 | 499 | |||
AK181470.1 | - | 1 | 108 | |||
BC010601.1 | - | 5 | 502 | 3257153 | ||
BC119437.1 | - | 3 | 239 | |||
BC148274.1 | - | 3 | 239 |