Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trabd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026485.2 | NP_080761.2 | 7 | 728 | ||
GenBank mRNA | AK005300.1 | - | 7 | 698 | ||
AK017745.1 | - | 7 | 722 | |||
AK131116.1 | - | 1 | 2461 | |||
AK146027.1 | - | 7 | 744 | |||
AK185880.1 | - | 3 | 288 | |||
BC005574.1 | - | 7 | 754 | 3967961 | ||
BU510197.1 | - | 7 | 808 |