Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam134c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026501.2 | NP_080777.1 | 8 | 914 | ||
NM_028933.3 | NP_083209.1 | 7 | 806 | |||
GenBank mRNA | AK004960.1 | - | 8 | 930 | ||
AK004979.1 | - | 8 | 917 | |||
AK016648.1 | - | 10 | 1087 | |||
AK051025.1 | - | 7 | 807 | |||
AK151685.1 | - | 8 | 915 | |||
AK160000.1 | - | 8 | 882 | |||
AK163482.1 | - | 8 | 920 | |||
AK180251.1 | - | 2 | 116 | |||
AK192965.1 | - | 2 | 116 | |||
AK207521.1 | - | 2 | 93 | |||
BC006712.1 | - | 5 | 504 | |||
BC016089.1 | - | 8 | 887 | 4503475 |