Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197980.1 | NP_932097.1 | 2 | 130 | ||
GenBank mRNA | AK002486.1 | - | 2 | 146 | ||
AK003697.1 | - | 2 | 174 | |||
AK007984.1 | - | 2 | 138 | |||
AK013260.1 | - | 2 | 146 | |||
AK013289.1 | - | 2 | 174 | |||
AK148526.1 | - | 2 | 103 | |||
BC031792.1 | - | 2 | 130 | 4193998 |