Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133693.2 | NP_598454.2 | 6 | 806 | ||
GenBank mRNA | AK005626.1 | - | 3 | 243 | ||
AK013929.1 | - | 6 | 747 | |||
AK151000.1 | - | 6 | 718 | |||
AK209948.1 | - | 3 | 195 | |||
AK210537.1 | - | 4 | 325 | |||
BC009148.1 | - | 6 | 743 | 3600406 | ||
BC011286.1 | - | 6 | 725 | 3485410 | ||
BC027372.1 | - | 6 | 730 | 4952304 | ||
BC039917.1 | - | 6 | 741 | |||
BC057928.1 | - | 6 | 732 | 5289543 |