Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142681.1 | NP_001136153.1 | 6, 7 | 671 | ||
NM_026522.5 | NP_080798.2 | 6, 7 | 671 | |||
GenBank mRNA | AK014071.1 | - | 6, 7 | 620 | ||
AK036717.1 | - | 6, 7 | 615 | |||
AK050655.1 | - | 6, 7 | 619 | |||
AK160156.1 | - | 6, 7 | 631 | |||
AK160200.1 | - | 6, 7 | 631 | |||
AK193697.1 | - | 1, 2 | 57 | |||
BC006876.1 | - | 6, 7 | 620 | 2647540 | ||
BC061063.1 | - | 6, 7 | 631 | 6591338 |