Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142681.1 | NP_001136153.1 | 12 | 1190 | ||
NM_026522.5 | NP_080798.2 | 10 | 932 | |||
GenBank mRNA | AK014071.1 | - | 12 | 1139 | ||
AK036717.1 | - | 12 | 1135 | |||
AK050655.1 | - | 10 | 880 | |||
AK147377.1 | - | 10 | 5988 | |||
AK160156.1 | - | 12 | 1150 | |||
AK160200.1 | - | 12 | 1151 | |||
BC006876.1 | - | 12 | 1139 | 2647540 | ||
BC061063.1 | - | 12 | 1150 | 6591338 |