Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291280.1 | NP_001278209.1 | 2 | 375 | ||
NM_026523.4 | NP_080799.1 | 2 | 375 | |||
GenBank mRNA | AA671747.1 | - | 2 | 346 | ||
AK011929.1 | - | 2 | 332 | |||
AK014077.1 | - | 2 | 354 | |||
BC028490.1 | - | 2 | 310 | 1366735 | ||
CB058776.1 | - | 2 | 387 |