Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp593 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024215.2 | NP_077177.2 | 1 | 132 | ||
GenBank mRNA | AK005247.1 | - | 1 | 134 | ||
AK014081.1 | - | 1 | 83 | |||
AK133585.1 | - | 1 | 130 | |||
AK156931.1 | - | 1 | 67 | |||
BC003465.1 | - | 1 | 69 | 2648747 |
Gene Symbol: E130218I03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021371.1 | - | 5 | 542 | ||
AK087464.1 | - | 3 | 1317 | |||
BC083116.1 | - | 4 | 848 | |||
CO429420.1 | - | 5 | 570 |