Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp593 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024215.2 | NP_077177.2 | 1 | 170 | ||
GenBank mRNA | AK005247.1 | - | 1 | 172 | ||
AK014081.1 | - | 1 | 121 | |||
AK133585.1 | - | 1 | 168 | |||
AK156931.1 | - | 1 | 105 | |||
BC003465.1 | - | 1 | 107 | 2648747 |
Gene Symbol: E130218I03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087464.1 | - | 3 | 1279 | ||
BC083116.1 | - | 4 | 810 | |||
CO429420.1 | - | 5 | 532 |