Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aen | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026531.4 | NP_080807.3 | 2 | 200 | ||
GenBank mRNA | AK012555.1 | - | 2 | 185 | ||
AK031972.1 | - | 2 | 201 | |||
AK167658.1 | - | 2 | 159 | |||
AK188340.1 | - | 2 | 226 | |||
BC022614.1 | - | 2 | 280 | 4167800 |