Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lurap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026547.1 | NP_080823.1 | 2 | 550 | ||
GenBank mRNA | AK013564.1 | - | 2 | 550 | ||
BC147058.1 | - | 2 | 507 | 9056069 | ||
BC147059.1 | - | 2 | 507 | 9056037 | ||
BC147737.1 | - | 2 | 507 | 9056686 | ||
BC147750.1 | - | 2 | 507 | 9056699 |