Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170486.1 | NP_001163957.1 | 1, 2 | 106 | ||
NM_026552.3 | NP_080828.1 | 2, 3 | 225 | |||
GenBank mRNA | AJ278129.1 | - | 2, 3 | 151 | ||
AK017993.1 | - | 2, 3 | 226 | |||
AK018069.1 | - | 2, 3 | 163 | |||
AK030840.1 | - | 1 | 3808 | |||
AK039215.1 | - | 2, 3 | 173 | |||
AK076248.1 | - | 2, 3 | 173 | |||
AK077325.1 | - | 2, 3 | 147 | |||
AK133626.1 | - | 2, 3 | 173 | |||
AK136082.1 | - | 2, 3 | 168 | |||
AK150690.1 | - | 2, 3 | 159 | |||
AK153380.1 | - | 2, 3 | 157 | |||
AK159410.1 | - | 1, 2 | 54 | |||
AK166440.1 | - | 2, 3 | 147 | |||
AK193553.1 | - | 1, 2 | 96 | |||
AK194784.1 | - | 1, 2 | 96 | |||
AK197517.1 | - | 1, 2 | 96 | |||
BC015280.1 | - | 1, 2 | 137 | |||
BC055309.1 | - | 1, 2 | 136 | 6529255 |