Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170485.1 | NP_001163956.1 | 3 | 309 | ||
NM_001170486.1 | NP_001163957.1 | 4 | 421 | |||
NM_026552.3 | NP_080828.1 | 5 | 540 | |||
GenBank mRNA | AJ278129.1 | - | 5 | 466 | ||
AK017993.1 | - | 5 | 541 | |||
AK018069.1 | - | 5 | 478 | |||
AK039215.1 | - | 5 | 488 | |||
AK076248.1 | - | 5 | 488 | |||
AK077325.1 | - | 5 | 462 | |||
AK133626.1 | - | 5 | 488 | |||
AK136082.1 | - | 5 | 483 | |||
AK150690.1 | - | 5 | 474 | |||
AK153380.1 | - | 5 | 472 | |||
AK159410.1 | - | 4 | 369 | |||
AK166440.1 | - | 5 | 462 | |||
BC015280.1 | - | 4 | 452 | |||
BC055309.1 | - | 4 | 451 | 6529255 |