Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdccag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085407.1 | NP_001078876.1 | 6, 7 | 947 | ||
NM_001085408.1 | NP_001078877.1 | 5, 6 | 878 | |||
NM_026563.3 | NP_080839.2 | 7, 8 | 1097 | |||
GenBank mRNA | AK014885.1 | - | 7, 8 | 1066 | ||
AK016227.1 | - | 1, 2 | 41 | |||
AK082787.1 | - | 6, 7 | 918 | |||
AK087227.1 | - | 1, 2 | 463 | |||
AK163690.1 | - | 5, 6 | 849 | |||
BC031199.1 | - | 5, 6 | 827 | 5064948 |