Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fdx1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039824.2 | NP_001034913.1 | 1 | 106 | ||
GenBank mRNA | AK008401.2 | - | 1 | 104 | ||
AK010211.1 | - | 1 | 77 | |||
AK020979.1 | - | 1 | 110 | |||
BC061189.1 | - | 1 | 123 | |||
BC172154.1 | - | 1 | 206 |