Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam212a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026597.3 | NP_080873.2 | 2 | 644 | ||
GenBank mRNA | AK020090.1 | - | 2 | 608 | ||
AK076092.1 | - | 2 | 608 | |||
BC051538.1 | - | 1 | 385 | |||
BC115538.1 | - | 2 | 583 | 40101503 | ||
BQ176112.1 | - | 1 | 430 |