Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sympk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026605.2 | NP_080881.2 | 16 | 2285 | ||
GenBank mRNA | AK019497.1 | - | 11 | 1761 | ||
AK146894.1 | - | 16 | 2266 | |||
AK155093.1 | - | 16 | 2286 | |||
AK155375.1 | - | 16 | 2240 | |||
AK165591.1 | - | 16 | 2264 | |||
AK168541.1 | - | 16 | 2266 | |||
AK168757.1 | - | 16 | 2266 | |||
AK172040.1 | - | 17 | 2314 | |||
BC049852.1 | - | 16 | 2230 | 4163760 |