Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024219.1 | NP_077181.1 | 1, 2 | 86 | ||
GenBank mRNA | AK010939.1 | - | 640 | |||
AK018708.1 | - | 1, 2 | 118 | |||
AK050550.1 | - | 1, 2 | 136 | |||
AK078498.1 | - | 1, 2 | 118 | |||
AK145940.1 | - | 1, 2 | 96 | |||
BC002153.1 | - | 1, 2 | 86 | 3485301 |