Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026614.2 | NP_080890.1 | 3 | 259 | ||
GenBank mRNA | AK008155.1 | - | 3 | 130 | ||
AK008156.1 | - | 3 | 125 | |||
AK008286.1 | - | 3 | 125 | |||
AK008414.1 | - | 3 | 125 | |||
AK009338.1 | - | 3 | 143 | |||
AK009396.1 | - | 3 | 129 | |||
AK009461.1 | - | 3 | 129 | |||
AK009493.1 | - | 3 | 128 | |||
AK009608.1 | - | 3 | 129 | |||
AK009642.1 | - | 3 | 144 | |||
AK009652.1 | - | 3 | 129 | |||
AK009775.1 | - | 3 | 153 | |||
AK009840.1 | - | 3 | 143 | |||
AK009985.1 | - | 3 | 143 | |||
AK010024.1 | - | 3 | 128 | |||
AK010090.1 | - | 3 | 131 | |||
AK010092.1 | - | 3 | 145 | |||
AK011579.1 | - | 3 | 119 | |||
AK011947.1 | - | 3 | 119 | |||
AK019084.1 | - | 3 | 130 | |||
AK019201.1 | - | 3 | 130 | |||
AK019207.1 | - | 3 | 121 | |||
AK019215.1 | - | 3 | 129 | |||
AK019242.1 | - | 3 | 127 | |||
AK019245.1 | - | 3 | 150 | |||
AK019291.1 | - | 3 | 130 | |||
AK019301.1 | - | 3 | 260 | |||
AK019324.1 | - | 3 | 239 | |||
AK019354.1 | - | 3 | 132 | |||
AK019359.1 | - | 3 | 151 | |||
AK019364.1 | - | 3 | 145 | |||
AK019368.1 | - | 3 | 143 | |||
AK019430.1 | - | 3 | 141 | |||
BC028633.1 | - | 3 | 127 | 1247576 |