Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026614.2 | NP_080890.1 | 5 | 476 | ||
GenBank mRNA | AK008155.1 | - | 5 | 346 | ||
AK008156.1 | - | 5 | 342 | |||
AK008286.1 | - | 5 | 342 | |||
AK008414.1 | - | 5 | 342 | |||
AK009338.1 | - | 5 | 360 | |||
AK009396.1 | - | 5 | 346 | |||
AK009461.1 | - | 5 | 346 | |||
AK009493.1 | - | 5 | 345 | |||
AK009608.1 | - | 5 | 346 | |||
AK009652.1 | - | 5 | 346 | |||
AK009775.1 | - | 5 | 370 | |||
AK009840.1 | - | 5 | 360 | |||
AK009985.1 | - | 5 | 360 | |||
AK010024.1 | - | 5 | 345 | |||
AK010090.1 | - | 5 | 348 | |||
AK010092.1 | - | 5 | 362 | |||
AK011579.1 | - | 5 | 336 | |||
AK011947.1 | - | 5 | 336 | |||
AK019084.1 | - | 5 | 347 | |||
AK019201.1 | - | 5 | 347 | |||
AK019207.1 | - | 5 | 338 | |||
AK019215.1 | - | 5 | 346 | |||
AK019242.1 | - | 5 | 344 | |||
AK019245.1 | - | 5 | 367 | |||
AK019291.1 | - | 5 | 347 | |||
AK019301.1 | - | 5 | 477 | |||
AK019324.1 | - | 5 | 456 | |||
AK019354.1 | - | 5 | 349 | |||
AK019359.1 | - | 5 | 368 | |||
AK019364.1 | - | 5 | 362 | |||
AK019368.1 | - | 5 | 360 | |||
AK019430.1 | - | 5 | 358 | |||
BC028633.1 | - | 5 | 344 | 1247576 |