Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930430F08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175128.2 | NP_780337.2 | 2 | 297 | ||
GenBank mRNA | AK015251.1 | - | 2 | 246 | ||
AK016270.1 | - | 129 | ||||
AK032226.1 | - | 2 | 253 | |||
AK036647.1 | - | 2 | 280 | |||
AK043358.1 | - | 1 | 2922 | |||
AK077761.1 | - | 2 | 291 | |||
AK078231.1 | - | 2 | 298 | |||
AK082894.1 | - | 2 | 250 | |||
AK160693.1 | - | 2 | 280 | |||
BC082587.1 | - | 2 | 247 | 30630155 | ||
CJ178338.1 | - | 2 | 298 |