Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 0610030E20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026696.1 | NP_080972.1 | 2 | 287 | ||
GenBank mRNA | AK002701.1 | - | 2 | 233 | ||
AK013079.2 | - | 1 | 343 | |||
AK085662.1 | - | 1 | 365 | |||
BB844069.1 | - | 2 | 287 | |||
BC064080.1 | - | 2 | 270 | 1246433 | ||
BC066053.1 | - | 2 | 261 | 6827710 | ||
BC117811.1 | - | 1 | 314 | |||
BC117812.1 | - | 2 | 237 | 40092656 |