Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291235.1 | NP_001278164.1 | 2 | 146 | ||
NM_001291236.1 | NP_001278165.1 | 2 | 622 | |||
NM_001291237.1 | NP_001278166.1 | 3 | 330 | |||
NM_026708.2 | NP_080984.1 | 2 | 457 | |||
GenBank mRNA | AA671983.1 | - | 3 | 347 | ||
AK002265.1 | - | 2 | 405 | |||
AK002703.1 | - | 2 | 353 | |||
AK046577.1 | - | 3 | 2337 | |||
AK161721.1 | - | 357 | ||||
AK165432.1 | - | 569 | ||||
BC005702.1 | - | 2 | 349 | 3598444 | ||
BC039182.1 | - | 2 | 592 | 3500908 | ||
BG083251.2 | - | 2 | 204 |