Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023455.3 | NP_075944.1 | 2 | 824 | ||
GenBank mRNA | AF163317.1 | - | 2 | 781 | ||
AK002784.1 | - | 2 | 765 | |||
BC019517.1 | - | 2 | 1016 | 4236543 |