Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026746.3 | NP_081022.2 | 5 | 634 | ||
GenBank mRNA | AK003685.1 | - | 5 | 586 | ||
BC009125.1 | - | 5 | 545 | 3155917 | ||
DV057668.1 | - | 5 | 654 |