Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144855.1 | NP_001138327.1 | 2, 3 | 738 | ||
GenBank mRNA | AK173053.1 | - | 2, 3 | 291 | ||
EU568872.1 | - | 2, 3 | 738 |