Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myadml2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026751.1 | NP_081027.1 | 1 | 732 | ||
GenBank mRNA | BC091000.1 | - | 1 | 505 | 6756293 | |
BC109329.1 | - | 1 | 471 | 40051378 | ||
BC125008.1 | - | 1 | 471 | 40051379 |