Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172400.3 | NP_765988.2 | 7 | 541 | ||
GenBank mRNA | AK003892.2 | - | 7 | 530 | ||
AK008163.1 | - | 543 | ||||
AK049526.1 | - | 6 | 1068 | |||
AK088325.1 | - | 7 | 545 | |||
AK150933.1 | - | 7 | 536 | |||
AK168645.1 | - | 7 | 525 | |||
BC035539.1 | - | 7 | 500 | |||
BC057191.1 | - | 7 | 512 | |||
BC064732.1 | - | 7 | 520 | |||
BC066222.1 | - | 7 | 520 | 30459247 | ||
BC080760.1 | - | 5 | 2937 | |||
BC137735.1 | - | 7 | 537 | 8860754 | ||
BC145655.1 | - | 7 | 537 | |||
BY754536.1 | - | 7 | 545 |