Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172400.3 | NP_765988.2 | 1 | 86 | ||
GenBank mRNA | AK003892.2 | - | 1 | 76 | ||
AK049526.1 | - | 1 | 79 | |||
AK088325.1 | - | 1 | 90 | |||
AK150933.1 | - | 1 | 81 | |||
AK168645.1 | - | 1 | 70 | |||
BC035539.1 | - | 1 | 45 | |||
BC057191.1 | - | 1 | 57 | |||
BC064732.1 | - | 1 | 65 | |||
BC066222.1 | - | 1 | 65 | 30459247 | ||
BC080760.1 | - | 1 | 65 | |||
BC137735.1 | - | 1 | 82 | 8860754 | ||
BC145655.1 | - | 1 | 82 | |||
BY754536.1 | - | 1 | 90 |