Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023479.2 | NP_075968.2 | 14, 15 | 1364 | ||
GenBank mRNA | AF308696.2 | - | 14, 15 | 1302 | ||
AK004136.1 | - | 2, 3 | 100 | |||
AK156730.1 | - | 14, 15 | 1365 | |||
AK172604.1 | - | 12, 13 | 1812 | |||
AK212327.1 | - | 1, 2 | 20 | |||
BC023134.1 | - | 14, 15 | 1303 | |||
BC048235.1 | - | 14, 15 | 1298 | 6493124 |