Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110271.1 | NP_001103741.1 | 6 | 1143 | ||
NM_001110272.1 | NP_001103742.1 | 5 | 888 | |||
NM_145919.2 | NP_666031.2 | 7 | 1019 | |||
GenBank mRNA | AI414039.1 | - | 1 | 236 | ||
AK003653.1 | - | 6 | 1028 | |||
CB055425.1 | - | 2 | 327 |