Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samm50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178614.4 | NP_848729.1 | 14 | 1387 | ||
GenBank mRNA | AK003990.1 | - | 14 | 1342 | ||
AK039949.1 | - | 14 | 1387 | |||
AK041783.1 | - | 1381 | ||||
AK084395.1 | - | 14 | 1381 | |||
AK086633.1 | - | 1383 | ||||
AK144975.1 | - | 14 | 1388 | |||
AK159740.1 | - | 14 | 1399 | |||
AK161378.1 | - | 14 | 1377 | |||
AK167806.1 | - | 14 | 1381 | |||
AK168510.1 | - | 14 | 1343 | |||
AK169066.1 | - | 14 | 1377 | |||
AK169351.1 | - | 14 | 1344 | |||
BC119178.1 | - | 14 | 1381 | 8733927 | ||
BC119180.1 | - | 14 | 1381 | 8733929 |