Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A730087J23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035979.1 | - | 1 | 342 | ||
AK043346.1 | - | 1 | 363 | |||
AK046904.1 | - | 1 | 315 |
Gene Symbol: Fam172a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163419.1 | NP_001156891.1 | 10 | 1212 | ||
NM_001163420.1 | NP_001156892.1 | 9 | 1020 | |||
NM_138312.1 | NP_612185.1 | 11 | 1336 | |||
GenBank mRNA | AK052419.1 | - | 10 | 1185 | ||
AK165273.1 | - | 11 | 1235 | |||
AK166285.1 | - | 9 | 992 | |||
BC057380.1 | - | 10 | 1180 |