Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smtnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024230.2 | NP_077192.1 | 2 | 600 | ||
GenBank mRNA | AK003989.1 | - | 2 | 502 | ||
BC002317.1 | - | 2 | 600 | 3593616 |