Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc44a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199186.1 | NP_001186115.1 | 12 | 1083 | ||
NM_152808.3 | NP_690021.1 | 12 | 1085 | |||
GenBank mRNA | AK036249.1 | - | 1 | 443 | ||
AK041533.1 | - | 12 | 1042 | |||
AK048648.1 | - | 12 | 1070 | |||
AK153992.1 | - | 12 | 1064 | |||
AK166695.1 | - | 12 | 1086 | |||
BC013213.1 | - | 5 | 478 | |||
BC031535.1 | - | 12 | 1061 | 3991609 |