Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026814.3 | NP_081090.1 | 1 | 169 | ||
GenBank mRNA | AK004064.1 | - | 1 | 170 | ||
BC115957.1 | - | 1 | 150 | 40055543 |