Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026836.2 | NP_081112.1 | 1 | 186 | ||
GenBank mRNA | AK050689.1 | - | 1 | 187 | ||
AK133851.1 | - | 2 | 1170 | |||
BC005603.1 | - | 1 | 165 | 3483315 |