Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134255.3 | NP_599016.2 | 5 | 590 | ||
GenBank mRNA | AK028761.1 | - | 5 | 530 | ||
AK045274.1 | - | 5 | 569 | |||
AK051580.1 | - | 5 | 587 | |||
AK085663.1 | - | 5 | 553 | |||
AK085696.1 | - | 5 | 540 | |||
AK088153.1 | - | 5 | 528 | |||
AK162451.1 | - | 5 | 591 | |||
AK167755.1 | - | 5 | 530 | |||
AK168360.1 | - | 5 | 530 | |||
AK168987.1 | - | 5 | 517 | |||
AK214227.1 | - | 1 | 90 | |||
BC022911.1 | - | 5 | 499 | 4457285 | ||
KF987081.1 | - | 5 | 473 |