Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134255.3 | NP_599016.2 | 7 | 869 | ||
GenBank mRNA | AK028761.1 | - | 7 | 809 | ||
AK031743.1 | - | 7 | 872 | |||
AK045274.1 | - | 7 | 848 | |||
AK051580.1 | - | 7 | 866 | |||
AK085663.1 | - | 7 | 832 | |||
AK085696.1 | - | 7 | 819 | |||
AK088153.1 | - | 7 | 807 | |||
AK162451.1 | - | 7 | 870 | |||
AK167755.1 | - | 7 | 809 | |||
AK168360.1 | - | 7 | 809 | |||
AK168987.1 | - | 7 | 796 | |||
BC022911.1 | - | 7 | 778 | 4457285 | ||
KF987081.1 | - | 7 | 752 |