Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mypn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182992.2 | NP_892037.2 | 6, 7 | 1541 | ||
GenBank mRNA | AK036458.1 | - | 6, 7 | 1502 | ||
AK220521.1 | - | 6, 7 | 1541 |